AMBH3155F905 Ambeed, Inc.

cis-3-Hexenyl acetate

  • MDL number MFCD00036563
  • Cas Number : 3681-71-8
  • Molecular Weight : 142.2000

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form liquid
Shipping Temperature Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/3681-71-8.html

Safety Information

signalword Danger
pcodes P210-P233-P240-P241+P242+P243-P280-P303+P361+P353-P370+P378-P403+P235-P501
MSDS https://www.ambeed.com/products/3681-71-8.html
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart