AMBH3155F92D Ambeed, Inc.

2-Aminoacetonitrile sulfate(2:1)

  • MDL number MFCD01941514
  • Cas Number : 5466-22-8
  • Molecular Weight : 210.2116

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/5466-22-8.html

Safety Information

signalword Danger
pcodes P264-P270-P280-P301+P310+P330-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313-P405-P501
MSDS https://www.ambeed.com/products/5466-22-8.html
pictograms GHS06
hstatements H301-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart