AMBH3155F930 Ambeed, Inc.

2,6-Bis((S)-4-isopropyl-4,5-dihydrooxazol-2-yl)pyridine

  • MDL number MFCD00191815
  • Cas Number : 118949-61-4
  • Molecular Weight : 301.3800

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/118949-61-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/118949-61-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart