AMBH3155F96A Ambeed, Inc.

9,9'-Spirobifluorene-2-boronic Acid

  • MDL number MFCD13248568
  • Cas Number : 236389-21-2
  • Molecular Weight : 360.2123

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/236389-21-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/236389-21-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart