AMBH3155F973 Ambeed, Inc.

6-Bromohexan-2-one

  • MDL number MFCD00044891
  • Cas Number : 10226-29-6
  • Molecular Weight : 179.0600

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form Colorless to light-yellow to brown powder or crystals or liquid
Shipping Temperature Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/10226-29-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/10226-29-6.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart