AMBH3155F991 Ambeed, Inc.

N-Benzylacrylamide

  • MDL number MFCD00015333
  • Cas Number : 13304-62-6
  • Molecular Weight : 161.2004

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/13304-62-6.html

Safety Information

signalword Danger
pcodes P261-P280-P301+P310-P305+P351+P338-P405-P501
MSDS https://www.ambeed.com/products/13304-62-6.html
pictograms GHS06
hstatements H301-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart