AMBH3155F99C Ambeed, Inc.

3-(Difluoromethoxy)-4-fluorobenzaldehyde

  • MDL number MFCD14698501
  • Cas Number : 1214367-20-0
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1214367-20-0.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1214367-20-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart