AMBH3155F99D Ambeed, Inc.

(Z)-Hex-3-en-1-yl 2-methylbutanoate

  • MDL number MFCD00036530
  • Cas Number : 53398-85-9
  • Molecular Weight : 184.2800

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/53398-85-9.html

Safety Information

signalword Danger
pcodes P273
MSDS https://www.ambeed.com/products/53398-85-9.html
pictograms GHS09
hstatements H411
Availability details for
Quantity
  • Availability
  • Add to Cart