AMBH3155F9A5 Ambeed, Inc.

Bis(lauroyloxy)dioctyltin

  • MDL number MFCD00026557
  • Cas Number : 3648-18-8
  • Molecular Weight : 745.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 96
Storage temp. Sealed in dry,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/3648-18-8.html

Safety Information

signalword Warning
pcodes P273
MSDS https://www.ambeed.com/products/3648-18-8.html
pictograms GHS07
hstatements H302-H332-H413
Availability details for
Quantity
  • Availability
  • Add to Cart