AMBH3155F9B4 Ambeed, Inc.

4-[[(4-Fluorophenyl)imino]methyl]phenol

  • MDL number MFCD00029739
  • Cas Number : 3382-63-6
  • Molecular Weight : 215.2200

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/3382-63-6.html

Safety Information

signalword Danger
pcodes P201-P261-P305+P351+P338-P308+P313
MSDS https://www.ambeed.com/products/3382-63-6.html
pictograms GHS07,GHS08
hstatements H302-H315-H319-H335-H350-H360
Availability details for
Quantity
  • Availability
  • Add to Cart