AMBH3155F9B9 Ambeed, Inc.

2-(3-Fluoro-4-nitrophenyl)acetic acid

  • MDL number MFCD03094239
  • Cas Number : 163395-24-2
  • Molecular Weight : 199.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 186.5°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 384.8°27.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/163395-24-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/163395-24-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart