AMBH3155F9C3 Ambeed, Inc.

2-Propoxyethanol

  • MDL number MFCD00072645
  • Cas Number : 2807-30-9
  • Molecular Weight : 104.1476

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2807-30-9.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/2807-30-9.html
pictograms GHS02,GHS07
hstatements H226-H312-H319
Availability details for
Quantity
  • Availability
  • Add to Cart