AMBH3155F9CA Ambeed, Inc.

Diethyl 2-(sec-butyl)malonate

  • MDL number MFCD00015194
  • Cas Number : 83-27-2
  • Molecular Weight : 216.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225°C at N/A mmHg
Purity 95
Flash Point 103.1°C
Storage temp. Sealed in dry,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 247.5°C at 760 mmHg
Product Link https://www.ambeed.com/products/83-27-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/83-27-2.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart