AMBH3155F9D3 Ambeed, Inc.

3-Bromo-2,6-dimethoxypyridine

  • MDL number MFCD08669938
  • Cas Number : 13445-16-4
  • Molecular Weight : 218.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 89°C
Storage temp. Inert atmosphere,Room Temperature
Physical Form Colorless or White to yellow powder or crystals or Liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 223.6°C at 760 mmHg
Product Link https://www.ambeed.com/products/13445-16-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13445-16-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart