AMBH3155F9DB Ambeed, Inc.

malonic acid dibutyl ester

  • MDL number MFCD00053795
  • Cas Number : 1190-39-2
  • Molecular Weight : 216.2741

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1190-39-2.html

Safety Information

signalword Danger
pcodes P210-P280-P370+P378-P403+P235-P501
MSDS https://www.ambeed.com/products/1190-39-2.html
pictograms GHS02
hstatements H227
Availability details for
Quantity
  • Availability
  • Add to Cart