AMBH3155F9DF Ambeed, Inc.

Cerium(III) oxalate xhydrate

  • MDL number N/A
  • Cas Number : 15750-47-7
  • Molecular Weight : 568.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/15750-47-7.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/15750-47-7.html
pictograms GHS07
hstatements H302+H312
Availability details for
Quantity
  • Availability
  • Add to Cart