AMBH3155F9EE Ambeed, Inc.

Phthaloylamlodipine

  • MDL number MFCD07782096
  • Cas Number : 88150-62-3
  • Molecular Weight : 538.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-145°C at N/A mmHg
Synonym 3-Ethyl 5-methyl 4-(2-chlorophenyl)-2-((2-(1,3-dioxoisoindolin-2-yl)ethoxy)methyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Purity 95
Flash Point 349.364°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 654.061°C at 760 mmHg
Product Link https://www.ambeed.com/products/88150-62-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/88150-62-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart