AMBH3155F9EF Ambeed, Inc.

3-Bromo-5-(prop-1-en-2-yl)pyridine

  • MDL number MFCD19689077
  • Cas Number : 118775-69-2
  • Molecular Weight : 198.0600

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/118775-69-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/118775-69-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart