AMBH3155F9FA Ambeed, Inc.

Methyl 4-dimethylaminobenzoate

  • MDL number MFCD00014905
  • Cas Number : 1202-25-1
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-102°C at N/A mmHg
Purity 98
Flash Point 112.6°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 280.3°C at 760 mmHg
Product Link https://www.ambeed.com/products/1202-25-1.html

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/1202-25-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart