AMBH3155FA03 Ambeed, Inc.

2-(3,4-Dihydro-2H-pyran-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD13195758
  • Cas Number : 1046811-99-7
  • Molecular Weight : 210.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Flash Point 88.3°C
Storage temp. Inert atmosphere,Store in freezer, under -20°C
Physical Form liquid
Shipping Temperature Shipped in cold pack
Country of Origin China
Boiling point 222.3°50.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/1046811-99-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1046811-99-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart