AMBH3155FA08 Ambeed, Inc.

2-Butyrylpyridine

  • MDL number MFCD00778319
  • Cas Number : 22971-32-0
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 98.6°C
Storage temp. Inert atmosphere,Room Temperature
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 230.4°C at 760 mmHg
Product Link https://www.ambeed.com/products/22971-32-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/22971-32-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart