AMBH3155FA09 Ambeed, Inc.

5-(Trifluoromethyl)-3,4-dihydronaphthalen-1(2H)-one

  • MDL number MFCD08234372
  • Cas Number : 885268-02-0
  • Molecular Weight : 214.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 113°C
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 257.6°40.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/885268-02-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/885268-02-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart