AMBH3155FA10 Ambeed, Inc.

4-Chloro-6-iodo-5-methylthieno[2,3-d]pyrimidine

  • MDL number MFCD26517433
  • Cas Number : 1823372-30-0
  • Molecular Weight : 310.5400

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1823372-30-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1823372-30-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart