AMBH3155FA11 Ambeed, Inc.

4-[(Trimethylsilyl)ethynyl]pyridine

  • MDL number MFCD09258874
  • Cas Number : 133810-35-2
  • Molecular Weight : 175.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 85.6°C
Storage temp. Inert atmosphere,Room Temperature
Physical Form Colorless or White to yellow powder or crystals or Liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 218.0°22.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/133810-35-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/133810-35-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart