AMBH3155FA23 Ambeed, Inc.

Methyl 2-(2-Pyrimidyl)acetate

  • MDL number MFCD12198887
  • Cas Number : 60561-50-4
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 86.9°C
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 220.1°C at 760 mmHg
Product Link https://www.ambeed.com/products/60561-50-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/60561-50-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart