AMBH3155FA33 Ambeed, Inc.

1-Hydroxy-2-naphthaldehyde

  • MDL number MFCD00092325
  • Cas Number : 574-96-9
  • Molecular Weight : 172.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56°C at N/A mmHg
Purity 98
Flash Point 134.3°C
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 317°C at 760 mmHg
Product Link https://www.ambeed.com/products/574-96-9.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/574-96-9.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart