AMBH3155FA3F Ambeed, Inc.

trans-3-Fluorocinnamic acid

  • MDL number MFCD00004383
  • Cas Number : 20595-30-6
  • Molecular Weight : 166.1500

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/20595-30-6.html

Safety Information

signalword Danger
pcodes P261-P301+P310-P305+P351+P338
MSDS https://www.ambeed.com/products/20595-30-6.html
pictograms GHS06
hstatements H301-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart