AMBH3155FA42 Ambeed, Inc.

tert-Butyl 2,9-diazaspiro[5.5]undecane-2-carboxylate hydrochloride

  • MDL number MFCD13185055
  • Cas Number : 1023301-88-3
  • Molecular Weight : 290.8300

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1023301-88-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1023301-88-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart