AMBH3155FA4A Ambeed, Inc.

Praseodymium(III) chloride

  • MDL number MFCD00011176
  • Cas Number : 10361-79-2
  • Molecular Weight : 247.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 786°C at N/A mmHg
Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/10361-79-2.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/10361-79-2.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart