AMBH3155FA4E Ambeed, Inc.

2-(Pyridin-4-yl)benzaldehyde

  • MDL number MFCD02684098
  • Cas Number : 176526-00-4
  • Molecular Weight : 183.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 342.8°C at 760 mmHg
Product Link https://www.ambeed.com/products/176526-00-4.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/176526-00-4.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart