AMBH3155FA50 Ambeed, Inc.

1-Boc-1,8-diazaspiro[5.5]undecane

  • MDL number MFCD10700119
  • Cas Number : 960294-18-2
  • Molecular Weight : 254.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 168°C
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 354.1°15.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/960294-18-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/960294-18-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart