AMBH3155FA6D Ambeed, Inc.

(Z)-3,7-Dimethylocta-2,6-dien-1-yl acetate

  • MDL number MFCD00063205
  • Cas Number : 141-12-8
  • Molecular Weight : 196.2860

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/141-12-8.html

Safety Information

signalword Danger
pcodes P261-P280-P273-P305+P351+P338
MSDS https://www.ambeed.com/products/141-12-8.html
pictograms GHS07,GHS09
hstatements H315-H317-H319-H411
Availability details for
Quantity
  • Availability
  • Add to Cart