AMBH3155FA6F Ambeed, Inc.

(-)-Camphor

  • MDL number MFCD00064148
  • Cas Number : 464-48-2
  • Molecular Weight : 152.2334

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/464-48-2.html

Safety Information

signalword Danger
pcodes P210-P261-P305+P351+P338
MSDS https://www.ambeed.com/products/464-48-2.html
pictograms GHS02,GHS07
hstatements H228-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart