AMBH3155FA72 Ambeed, Inc.

4'-(Trifluoromethoxy)-5-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid

  • MDL number MFCD18322456
  • Cas Number : 1261503-00-7
  • Molecular Weight : 350.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4'-(trifluoromethoxy)-5-(trifluoromethyl)[1,1'-biphenyl]-3-carboxylic acid
Inchi Code 1S/C15H8F6O3/c16-14(17,18)11-6-9(5-10(7-11)13(22)23)8-1-3-12(4-2-8)24-15(19,20)21/h1-7H,(H,22,23)
Inchi Key CVCGIORMQQAACT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1261503-00-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1261503-00-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart