AMBH3155FA8D Ambeed, Inc.

Methyl 4-hydroxypiperidine-2-carboxylate

  • MDL number MFCD09152760
  • Cas Number : 144913-66-6
  • Molecular Weight : 159.1800

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form Colorless or white to yellow liquid or powder or crystals
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/144913-66-6.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/144913-66-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart