AMBH3155FA9D Ambeed, Inc.

4,4-Diamino-2,2-stilbenedisulfonic acid

  • MDL number MFCD00024946
  • Cas Number : 81-11-8
  • Molecular Weight : 370.4008

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/81-11-8.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/81-11-8.html
pictograms GHS05,GHS07
hstatements H302-H318
Availability details for
Quantity
  • Availability
  • Add to Cart