AMBH3155FAA8 Ambeed, Inc.

Sorbic acid

  • MDL number MFCD00002703
  • Cas Number : 110-44-1
  • Molecular Weight : 112.1300

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/110-44-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/110-44-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart