AMBH3155FAB2 Ambeed, Inc.

3-Fluoro-5-methoxyisonicotinaldehyde

  • MDL number MFCD18258153
  • Cas Number : 1256792-34-3
  • Molecular Weight : 155.1300

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1256792-34-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501
MSDS https://www.ambeed.com/products/1256792-34-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart