AMBH3155FAC3 Ambeed, Inc.

3-Ethyloxetan-3-ol

  • MDL number MFCD22690226
  • Cas Number : 1416438-41-9
  • Molecular Weight : 102.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 156.4��t 760 mmHg
Iupac Name 3-ethyl-3-oxetanol
Inchi Code 1S/C5H10O2/c1-2-5(6)3-7-4-5/h6H,2-4H2,1H3
Inchi Key QGXMKEGFTNLFQQ-UHFFFAOYSA-N

Description

Purity 97%
Flash Point 61.5
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Boiling point 156.4��t 760 mmHg
Product Link https://www.ambeed.com/products/1416438-41-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1416438-41-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart