AMBH3155FB31 Ambeed, Inc.

6-Bromo-3-methyl-2H-indazole

  • MDL number MFCD20488637
  • Cas Number : 90221-46-8
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-3-methyl-2H-indazole
Inchi Code 1S/C8H7BrN2/c1-5-7-3-2-6(9)4-8(7)11-10-5/h2-4H,1H3,(H,10,11)
Inchi Key PUVRYFUBGFMXMW-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/90221-46-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/90221-46-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart