AMBH3155FB4C Ambeed, Inc.

Ethyl sorbate

  • MDL number MFCD00009296
  • Cas Number : 2396-84-1
  • Molecular Weight : 140.1800

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2396-84-1.html

Safety Information

signalword Danger
pcodes P273
MSDS https://www.ambeed.com/products/2396-84-1.html
pictograms GHS09
hstatements H411
Availability details for
Quantity
  • Availability
  • Add to Cart