AMBH3155FB66 Ambeed, Inc.

Methyl 4-acetyl-3-nitrobenzoate

  • MDL number MFCD30189414
  • Cas Number : 51885-81-5
  • Molecular Weight : 223.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-acetyl-3-nitrobenzoate
Inchi Code 1S/C10H9NO5/c1-6(12)8-4-3-7(10(13)16-2)5-9(8)11(14)15/h3-5H,1-2H3
Inchi Key RWLGQQWVBIMMEG-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/51885-81-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/51885-81-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart