AMBH3155FB7E Ambeed, Inc.

3-Chloro-4-fluorobenzylamine

  • MDL number MFCD00042507
  • Cas Number : 72235-56-4
  • Molecular Weight : 159.5886

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/72235-56-4.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/72235-56-4.html
pictograms GHS05
hstatements H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart