AMBH3155FB87 Ambeed, Inc.

9-(tert-Butoxy)-9-oxononanoic acid

  • MDL number MFCD30737825
  • Cas Number : 1290540-34-9
  • Molecular Weight : 244.3273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form White to yellow powder or crystals or liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1290540-34-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1290540-34-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart