AMBH3155FBE1 Ambeed, Inc.

N,N-Diethyl-2-phenylacetamide

  • MDL number MFCD00192031
  • Cas Number : 2431-96-1
  • Molecular Weight : 191.2695

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form Colorless to yellow liquid or white to yellow powder or crystals
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2431-96-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2431-96-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart