AMBH3155FC2A Ambeed, Inc.

(E)-2-Methylpent-2-enoic acid

  • MDL number MFCD00002655
  • Cas Number : 16957-70-3
  • Molecular Weight : 114.1424

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,Room Temperature
Physical Form Solid or liquid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/16957-70-3.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/16957-70-3.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart