AMBH3155FC4E Ambeed, Inc.

6-Nitroquinoxaline-2,3-dione

  • MDL number MFCD00673833
  • Cas Number : 2379-56-8
  • Molecular Weight : 205.1300

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2379-56-8.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/2379-56-8.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart