AMBH3155FC5C Ambeed, Inc.

2-Nitro-4-trifluoromethyl-benzoyl chloride

  • MDL number MFCD07777203
  • Cas Number : 81108-81-8
  • Molecular Weight : 253.5600

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/81108-81-8.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/81108-81-8.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart