AMBH3155FC61 Ambeed, Inc.

(2R,4R)-Ethyl 4-methylpiperidine-2-carboxylate

  • MDL number MFCD03093749
  • Cas Number : 74892-82-3
  • Molecular Weight : 171.2400

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,Store in freezer, under -20
Physical Form liquid
Shipping Temperature Keep in dark place,Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/74892-82-3.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/74892-82-3.html
pictograms GHS07
hstatements H227-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart