AMBH3155FC68 Ambeed, Inc.

Cefcapene pivoxil hydrochloride hydrate

  • MDL number MFCD01682041
  • Cas Number : 147816-24-8
  • Molecular Weight : 622.1113

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 722764
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/147816-24-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/147816-24-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart